Learn More
Stoelting™ 1700 Series Gas-Tight Microliter Syringe
SDP
Catalog No. 10000910 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Bevel tip
Blunt tip
Luer tip
Size:
10 μL syringe
10 μL syringe; 26 ga
100 μL syringe
100 μL syringe; 22 ga
25 μL syringe
25 μL syringe; 22 ga
250 μL syringe
250 μL syringe; 22 ga
50 μL syringe
50 μL syringe; 22 ga
500 μL syringe
500 μL syringe; 22 ga
Includes:
Removable Needle
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Size Includes
10-000-910 Bevel tip 250 μL syringe; 22 ga Removable Needle
10-000-896 Bevel tip 10 μL syringe; 26 ga
10-000-901 Blunt tip 10 μL syringe; 26 ga
10-000-889 Luer tip 10 μL syringe
10-000-899 Bevel tip 100 μL syringe; 22 ga
10-000-909 Bevel tip 100 μL syringe; 22 ga Removable Needle
10-000-904 Blunt tip 100 μL syringe; 22 ga
10-000-915 Blunt tip 100 μL syringe; 22 ga Removable Needle
10-000-892 Luer tip 100 μL syringe
10-000-897 Bevel tip 25 μL syringe; 22 ga
10-000-902 Blunt tip 25 μL syringe; 22 ga
10-000-890 Luer tip 25 μL syringe
10-000-900 Bevel tip 250 μL syringe; 22 ga
10-000-905 Blunt tip 250 μL syringe; 22 ga
10-000-916 Blunt tip 250 μL syringe; 22 ga Removable Needle
10-000-893 Luer tip 250 μL syringe
10-000-903 Blunt tip 50 μL syringe; 22 ga
10-000-898 Bevel tip 50 μL syringe; 22 ga
10-000-891 Luer tip 50 μL syringe
10-000-917 Blunt tip 500 μL syringe; 22 ga Removable Needle
10-000-894 Luer tip 500 μL syringe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Catalog No. 10-000-910 Supplier Stoelting™ Supplier No. 53483
Only null left
Add to Cart
Add to Cart
A variety of different terminations and needle options are available in this series.
Ideal for dispensing volumes from 1 μL up to 500 μL.
Specifications
For Use With (Application) Neuroscience
Size 250 μL syringe; 22 ga
Model Bevel tip
Includes Removable Needle