Learn More
3M™ SecureFit™ 600 Series Protective Eyewear
Catalog No. 17000681 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Includes:
Foam gasket
Model:
SF601SGAF
SF601SGAF-FM
SF602SGAF
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Includes Model
17-000-681 Gray SF602SGAF
17-000-680 Clear SF601SGAF
17-000-682 Clear Foam gasket SF601SGAF-FM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-000-681 Supplier 3M™ Supplier No. 7100128690
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ideal eye protection for diverse workforces. The 3M™ SecureFit™ 600 Series Protective Eyewear features 3M Pressure Diffusion Temple Technology, which provides for a more secure and comfortable fit.
  • Features proprietary 3M Pressure Diffusion Temple Technology which helps diffuse pressure over the ear for a secure, comfortable fit
  • Scotchgard Anti-fog Coating lasts longer than traditional anti-fog coatings, helping to increase eyewear performance in wet and steamy environments
  • The proprietary coating retains its effectiveness for at least 25 washings, allowing workers to wear their eyewear longer
  • Workers can feel confident wearing Scotchgard Anti-Fog coated eyewear because it meets the requirements of ANSI Z87.1.2015
  • Clear polycarbonate lens absorbs 99.9% UVA & UVB
  • 3M Catalog# SF601SGAF , 3M Catalog# SF601SGAF-FM , 3M Catalog# SF602SGAF
Compliance
Meets ANSI Z87.1-2015 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Gray
For Use With (Application) Automotive, Construction, General Manufacturing, Metal Fabrication, Mining, Oil & Gas, Pharmaceutical, Primary Metals
Frame Color Gray
Includes
Model SF602SGAF
Temple Style SecureFit™
Lens Material Polycarbonate
Frame Material Plastic
Frame Type Half Frame