Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl N-Boc-Oxamidate

Catalog No. 173465G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-346-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 17-346-5G Supplier Sigma Aldrich Supplier No. 173465G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent for transforming alcohols into BOC-protected amines by Mitsunobu reaction (PPh3/DEAD); prim and sec alcohols (inversion of configuration) give good yields

Specifications

CAS 73522-42-6
Density 0.915 g/mL at 25°C
Boiling Point 94°C to 99°C27 mmHg, lit.)
Molecular Formula C10H19N
Linear Formula C10H19N
Quantity 5 g
MDL Number MFCD00001349
Molecular Weight (g/mol) 153.26 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.