Learn More
3M™ E-A-R™ UltraFit™ Metal Detectable Corded Ear Plugs
Catalog No. 173806 Shop All 3M Products
Change view
Click to view available options
400 Pair/Cs.
Catalog No.
17-380-6 400 Pair/Cs.
1 options
Catalog No. 17-380-6 Supplier 3M™ Supplier No. 7000002319
Add to Cart
Add to Cart
E-A-R™ metal detectable ear plugs have a stainless steel bead encased in the ear plug. Popular in food manufacturing industries, when contamination prevention is critical.
  • Designed with detectable metal components for food processing and other industries
  • Low-pressure in the ear canal for enhanced comfort
  • Poly bag keeps each pair of ear plugs clean before use
  • Moisture-resistant helps maintain a consistent expansion time, even in humid situations
  • Slightly longer than regular size 3M™ E-A-R™ Classic™ ear plugs for higher noise reduction
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
Cord Color Blue
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Food Processing, Paper and Pulp, Tobacco, and Other Various Industries That Require Detectable Products to Prevent Contamination
High Visibility No
Model 340-4007
Packaging Type Poly bag
Pairs per Package 400 Pairs/Cs
Plug Material Elastomeric polymer
Plug Color Blue
Noise Reduction Rating (NRR) 25 dB
Corded Corded
Plug Shape Flanged
Disposable Reusable
Metal Detectable Yes