Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA Professional Jeresette 301 FSC Certified Latex Gloves

Catalog No. 17468557 Shop All MAPA Professional Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-468-557 Medium
17-468-555 X-Small
17-468-556 Small
17-468-558 Large
17-468-559 X-Large
17-468-554 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-468-557 Supplier MAPA Professional Supplier No. 34301027
Add to Cart
Add to Cart

Offers sustainable hand protection made from FSC™ certified latex, designed for long-term use in tough work environments.

  • Provides maximum comfort and durability for long-term use in demanding environments.
  • Made from FSC™ certified latex, offering sustainable protection and eco-friendly performance.
  • Anatomically shaped with a seamless liner to ensure freedom of movement and all-day comfort.
  • High tear resistance for enhanced durability and extended glove lifespan.
  • Resistant to diluted acids and bases, delivering reliable chemical protection.
  • Silicone-free material prevents defects on glass and metal surfaces prior to painting.
  • Individually packaged with optimized materials, reducing plastic use and incorporating 30% recycled content.
  • Certified chemical protection: CAT. 3 0334 2131X Type B KPT, compliant with stringent safety standards.
  • This product is not classified as hazardous under Regulation (EC) No. 1272/2008 of the European Parliament and of the Council. It contains no more than 0.1% of substances of very high concern (SVHC) and no substances listed in Annex XVII of Regulation (EC) No. 1907/2006 (REACH). UE type certificate / CE type examination certificate: 0075/014/162/05/22/0905 EX 01/05/22.

Specifications

ANSI Abrasion Rating ANSI 2
Chlorinated No
Applications Industrial, Laboratory
Color Blue
Finger Thickness (English) 45 mil
Finger Thickness (Metric) 1.15 mm
Latex Free No
Material Latex
Palm Thickness (English) 45 mil
Palm Thickness (Metric) 1.15 mm
Product Type Chemical and Liquid Protection Gloves
Size Medium
Thickness (English) 45 mil
Thickness (Metric) 1.15 mm
EN388 Standard EN388: 2131x
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES B
Grip Design Pebble Embossed
Liner Material Textile Support
Lining Seamless Lining
Model Jersette 301
Length (English) 12 in.
Length (Metric) 30 cm
Product Line Jersette
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.