Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Disposable Protective Coveralls 4520, Collared

Catalog No. 17624105 Shop All 3M Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-624-105 Large
17-624-103 3X-Large
17-624-104 4X-Large
17-624-107 2X-Large
17-624-106 Medium
17-624-108 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-624-105 Supplier 3M™ Supplier No. 7000088930
Add to Cart
Add to Cart
This item is not returnable. View return policy

Coveralls protect against certain light liquid splashes and hazardous dusts while enhancing movement and comfort.

  • Designed for use in industrial workplaces as a physical barrier to help prevent skin contact from liquid splashes and dust in certain work environments
  • Seamless shoulder and sleeve tops help provide increased comfort and fewer potential entry points for contaminants
  • Meet ANSI 101-1996 size guidelines
  • Meets EN 340, Type 6 Splash - EN 13034/(EN ISO 17491-4), Type 5 Dust - EN ISO 13982-1, Anti-Static, EN 11492 and Nuclear - EN 1073 - 2
  • Microporous PE laminate
  • Breathable, SMS fabric back panel helps improve air ventilation and reduce heat build-up
  • Reinforcement gusset between the legs to help provide increased durability during bending and squatting
  • Elastic waist and ankles, knit cuffs, two-way zipper with sealable storm flap
  • Standard collar for use with respiratory equipment or where a hood may not be desired
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material Microporous Polyethylene Film
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Knit
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Resistance Dust, Liquid
Coating Anti-static
Collar Type Standard Fold
Certifications/Compliance ANSI 101-1996
Certifications/Compliance (Europe) CE Category III Type 5/6, EN 13034, EN-ISO 13982-1, EN 1149-1:2006, EN 1073
Description 3 Layers of Melt-blown Polypropylene in-between 2 layers of Spunbound Polypropylene
Latex Free No
Model 4520
Size (English) Chest 39 to 43 in.
Size (English) Inseam 32 in.
Size (English) Waist 51 in.
Size (Metric) Chest 99.1 to 109.2 cm
Size (Metric) Waist 129.54 cm
For Use With (Application) Insulation Installation, Asbestos and Lead Abatement, Coal Dust in Power Plants, Woodworking, Metal Polishing, Light-Duty Building Cleaning, General Powder Handling, General Industrial Clean-up and Maintenance, Machine or Vehicle Maintenance and Food Processing
Height (English) 69 to 71 in.
Height (Metric) 174 to 181 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.