Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Qualitative Fit-Test Kit

Catalog No. 1798593 Shop All 3M Products
Change view
Click to view available options
Product Type:
Bitter Fit Test Kit
Qualitative Fit Test Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-985-93 Qualitative Fit Test Kit
19-002-720 Bitter Fit Test Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Catalog No. 17-985-93 Supplier 3M™ Supplier No. 7000001898
Add to Cart
Add to Cart
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

This Qualitative Fit-Test Kit contains everything necessary to fit test any particulate respirator or gas/vapor respirator with a particulate prefilter.

  • Fit test kit for respirators.
  • Kit includes:
    • Hood
    • Collar Assembly
    • Two Nebulizers
    • Sensitivity Solution, 55 mL (1.9 oz.)
    • Fit Test Solution, 55 mL (1.9 oz.)
    • Laminated User Instructions (Multilingual)
    • Two Sets of Replacement Nebulizer Inserts
  • Optional training and fit test case includes fit-test kit materials as well as training materials (videos, posters and literature).
  • FT-10 uses a test solution of sodium saccharin that produces a sweet tasting spray.
  • FT-30 uses a test solution of denatonium benzoate to produce a very bitter taste.

Usage:

  • Works by spraying the solution into the hood environment.
  • If the wearer does not detect any taste, the fit test has been passed.

Order Info

Individual 1.9 oz. (55 mL) bottles of sensitivity solution or fit test solution also available.

Compliance

Meets the performance criteria for fit testing respirators under OSHA 29 CFR 1910.134, Appendix A.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Qualitative Fit Test Kit
Certifications/Compliance OSHA 29 CFR 1910.134, Appendix A
For Use With (Application) Respirator Fit Testing
Includes Laminated instruction booklet, sensitivity solution, fit test solution, two nebulizers, hood, collar assembly, two sets replacement nebulizer inserts
Model FT-10

For Occupational Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.