Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ 6000 Series Cartridges

Catalog No. 1798611A Shop All 3M Products
Change view
Click to view available options
Cartridge Type:
Acid Gases: Chlorine, Hydrogen Chrloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Sulfide
Ammonia, Methylamine
Mercury Vapor, Chlorine, Sulfur Dioxide
Organic Vapor
Organic Vapors, Acid Gases, Ammonia, Methlamine, Formaldehyde
Organic Vapors, Acid Gases, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Sulfide, Hydrogen Fluoride
Organic Vapors, Formaldehyde
Color:
Black
Black/Olive
Gray
Green
Olive
White
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type Color
17-986-11A Organic Vapor Black
17-986-11B Acid Gases: Chlorine, Hydrogen Chrloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Sulfide White
17-986-11C Organic Vapors, Acid Gases, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Sulfide, Hydrogen Fluoride Yellow
17-986-11D Ammonia, Methylamine Green
17-986-11E Organic Vapors, Formaldehyde Black/Olive
18-999-4566 Organic Vapors, Acid Gases, Ammonia, Methlamine, Formaldehyde Olive
19-070-340 Mercury Vapor, Chlorine, Sulfur Dioxide Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-986-11A Supplier 3M™ Supplier No. 7000051851
Add to Cart
Edge
Add to Cart

NIOSH-approved cartridges for use with 3M™ full and half facepiece respirators 6000, 7000 and FF-400 Series.

  • For use with 3M full and half facepiece respirators 6000, 7000 and FF-400 Series
  • Bayonet cartridge connection
  • NIOSH-approved for protection against a variety of gases, vapors, and particulate hazards*
  • For contaminants up to 10X the Permissible Exposure Limit (PEL) with half-facepiece respirators or 50X the PEL (when fit tested) with full-facepiece respirators or according to specific OSHA standards or applicable government regulations, whichever is lower
  • Not intended for use in atmospheres that are unknown or immediately dangerous to life or death (IDLH)
  • Do not exceed maximum use concentrations established by regulatory standards
  • These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product
  • Misuse may result in sickness or death

*Details found at individual product level according

TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Organic Vapor
Certifications (NIOSH) NIOSH-approved
Connection Type Bayonet
Color Black
Product Type Cartridge
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, machine operation, painting
For Use With (Equipment) 3M™ half and full facepiece 6000, 7000, ff-400 series respirators
Model 6001
Quantity 60/Cs.
Length (English) 4.2 in.
Length (Metric) 10.6 cm
Height (English) 3.4 in.
Height (Metric) 8.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.