Learn More
Market Lab Tube Racks for 17mm Tubes, White, 2pk
Supplier:  Market Lab 1882WH
Encompass_Preferred
The ML1882-WH is a white tube rack from MarketLab that holds 17mm diameter tubes. It has 18 holes and can hold 18 tubes up to 100mm long. The rack is made from polypropylene and has an external size of 5.5" x 3.3" x 1.4". It can withstand temperatures from -20°C to 121°C. The rack allows easy access to tubes for retrieval and maximizes storage space. It can be autoclaved for sterilization and is durable and chemically resistant. This test tube rack is a useful organizational tool for holding and arranging tubes in the lab.
Catalog No. NC2683924
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.