Learn More
Market Lab Tube Racks for 17mm Tubes
Supplier:  Market Lab 1882YL
Encompass_Preferred
The ML1882-YL tube rack from MarketLab holds up to 50 17mm diameter tubes in 5 rows of 10. It is made from yellow polypropylene and has numbered positions from 1 to 50. The rack measures 16.5" x 3.5" x 2" and weighs 1 lb. This durable, chemical resistant rack allows easy access to tubes for storage or transport. The open wire design allows visibility of tube contents. It can be cleaned easily and is autoclavable up to 250°F. This is a useful organizational tool for any laboratory working with 17mm tubes.
Catalog No. NC2683921
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.