Alfa Aesar™ 18-Crown-6, 99%

Manufacturer: Alfa Aesar™ A1124909

Quantity 10g

 View more versions of this product

Catalog No. AAA1124909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17455-13-9
    Molecular Formula C12H24O6
    Formula Weight 264.32
    MDL Number MFCD00005113
    Beilstein 1619616
    Merck Index 14,2602
    Synonym 1,4,7,10,13,16-Hexaoxacyclooctadecane
    Chemical Name or Material 18-Crown-6

    Specifications

    Formula Weight 264.32
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    If eye irritation persists: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Merck Index 14,2602
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H319
    Harmful if swallowed.
    Causes serious eye irritation.
    TSCA TSCA
    Molecular Formula C12H24O6
    MDL Number MFCD00005113
    Beilstein 1619616
    Synonym 1,4,7,10,13,16-Hexaoxacyclooctadecane
    Boiling Point 116°C (0.2mmHg)
    Melting Point 37° to 41°C
    Flash Point 109°C (228°F)
    Odor Odorless
    CAS 17455-13-9
    Quantity 10g
    RTECS Number MP4500000
    Chemical Name or Material 18-Crown-6
    Assay 99%

    Description

    Applications
    18-Crown-6 is used as an efficient phase transfer catalyst and as a complexing agent with a variety of small cation. It is involved in the synthesis of diaryl ethers, diaryl thioethers, and diarylamines mediated by potassium fluoride-alumina and 18-crown-6. It facilitates the solubility of potassium permanganate in benzene, which is used for oxidizing the organic compounds. It is used to accelerate various substitution reactions as well as enhances the power of nucleophiles such as potassium acetate. It is utilized in the alkylation reactions in the presence of potassium carbonate, N-alkylation of glutarimide and succinimide with dimethylcarbonate. The complex formed by its reaction with potassium cyanide acts as a catalyst in the cyanosilylation of aldehydes, ketones and quinines with trimethylsilyl cyanide (TMSCN).

    Solubility
    Soluble in water.

    Notes
    Hygroscopic. Incompatible with strong oxidizing agents and strong acids.

    GHS H Statement
    H302-H319
    Harmful if swallowed.
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    If eye irritation persists: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit