Learn More
Gibco™ 18% Ethanol

Description
Gibco process buffers are sterile-filtered, packaged in bioprocess containers, and ready to order. Sourcing process buffers can allow you to focus on additional critical steps in your biopharmaceutical processes.
Features
- Available in a range of pack sizes
- Established shelf life; set specifications
- Made to order
- Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
- Readily accessible documentation to help accelerate qualification
Benefits
- Helps ease time requirements for in-house buffer preparation
- Mitigates space constraints for equipment storage
- Streamlines in-house processes related to formulation, hydration, testing, and validation
- Addresses workflow needs for biologics development and manufacturing stages
Additional information
- Multi-compendial testing: compendial-grade or equivalent raw materials
- Primary packaging: Aegis 5-14 single-use bioprocess container (BPC)
- Secondary packaging: rigid shipping container with appropriate dunnage
- Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
Specifications
Specifications
| Chemical Name or Material | Raw Material: 200 Proof Ethanol |
| CAS | 64-17-5 |
| Quantity | 5 L |
| Packaging | Bioprocess Container in Secondary Packaging |
| Boiling Point | Information not available. |
| Molecular Weight (g/mol) | Raw Material: 46.07 |
| Percent Purity | Not Available |
| Grade | ACS/BP/EP/JP/USP |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.