Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Adapter for 18-place Rotor

Catalog No. 05414318 Shop All Eppendorf North America Products
Change view
Click to view available options
No. per Pack:
18/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
05-414-318 18/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 05-414-318 Supplier Eppendorf™ Supplier No. 5427708006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for use with Eppendorf Cryovials™ and HPLC Tubes

  • Adapter
  • For 1 HPLC vessel 1.5 mL, for Rotor F-45-18-17-Cryo
TRUSTED_SUSTAINABILITY

Specifications

Type Adapter
For Use With (Application) 1.5mL HPLC Vials
No. per Pack 18/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.