Learn More
EATON CORP 1800 WATT 6OUTLET LINE COND.
SDP
Supplier:  EATON CORP LC1800
Encompass_Preferred
Line Conditioner/AVR System; Tripp Lite; 1800 watt; 6 outlets; Cord Length: 6 ft.; 15A maximum capacity; Maintains regulated 120V nominal output over an input range of 89 to 147V
Catalog No. 50-863-704
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.