Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KleenGuard™ G60 Medium Duty Polyurethane-Coated Cut-Resistant Gloves

Catalog No. 18042925 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-042-925 X-Large
18-042-922 Small
18-042-923 Medium
18-042-924 Large
18-042-926 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 18-042-925 Supplier KleenGuard™ Supplier No. 13826
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

KleenGuard™ G60 Medium Duty Polyurethane-Coated Cut-Resistant Gloves are constructed of ultra-high molecular weight polyethylene fibers to enhance breathability, and provide ANSI Cut Level 3 protection.

  • Constructed of ultra-high molecular weight polyethylene (UHMWPE) fibers to provide breathability and protection
  • Polyurethane coated palm and fingers enhance dexterity and grip capabilities
  • Separate right and left gloves in a pair provide a snug fit
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material HPPE
Product Type Cut-resistant Gloves
Size X-Large
EN388 Standard EN 388
For Use With (Application) Assembly, plastics, light raw materials
Glove Cuff Style Knit
Model 13826
Series KleenGuard™ G60
Yarn Gauge 13
Length (English) 10.2 in.
Length (Metric) 26 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.