Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Ethyl-3-Methylimidazolium Methyl Sulfate

Catalog No. 1808650GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-086-50GF 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 18-086-50GF Supplier Sigma Aldrich Supplier No. 1808650GF
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Ethyl-3-methylimidazolium methyl sulfate [EMIM][MS] when added as an eluent modifier to the mobile phase results in better separation of the three positional isomers of benzoic acid by reversed-phase (RP) high-performance liquid chromatography (HPLC).[EMIM][MS] may be used as a solvent to synthesize gold nanoparticles via reduction of gold(III) chloride trihydrate [HAuCl4.3H2O] using glycerol as a reducing agent.

Specifications

MDL Number MFCD00135579
Quantity 10 g
Synonym p-Alkoxy-benzyl alcohol polymer-bound; p-Alkoxybenzyl alcohol resin; [4-(Hydroxymethyl)phenoxymethyl]polystyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.