Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ HMIG and NFR Dispenser Labels
GREENER_CHOICESDP

Catalog No. 18999786 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
HMIG Dispenser Label
NFR Dispenser Label
Quantity:
500/Pk.
750/Pk.
Size (English):
0.87 x 0.87 in.
0.87 x 2.87 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity Size (English)
18-999-786 NFR Dispenser Label 500/Pk. 0.87 x 2.87 in.
18-999-784 HMIG Dispenser Label 500/Pk. 0.87 x 2.87 in.
18-999-785 HMIG Dispenser Label 750/Pk. 0.87 x 0.87 in.
18-999-787 NFR Dispenser Label 750/Pk. 0.87 x 0.87 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-999-786
Add to Cart
Add to Cart

Keep workers safe by properly informing them of workplace hazards.

  • Rolls come in a dispenser pack for convenient storage and use
  • HMIG labels feature a write-on area for specific hazard information
  • 18-999-786 NFR labels have room to write important information
  • 18-999-787 labels feature an NFR legend: call attention to various chemical hazards throughout the workplace

Order Info

  • Come in a dispenser pack for convenient storage and use
  • Hazardous Materials Identification Guide labels come in packs of 500 and 750, feature a write-on area for specific hazard information
  • 18-999-786 National Fire Rating labels come in packs of 500 and feature room to write important information
  • 18-999-787 labels are available in packs of 750 and feature an NFR legend, calling attention to various chemical hazards throughout the workplace

Specifications

Color Assorted
Format Label
Product Type NFR Dispenser Label
For Use With (Application) Keep workers safe by properly informing them of workplace hazards
Quantity 500/Pk.
Size (English) 0.87 x 2.87 in.
Size (Metric) 0.02 x 0.079 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.