Learn More
Justrite™ 19 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Self-Closing Doors
GSA_VAGREENER_CHOICE
Catalog No. 19822337 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Light Neutral
Silver
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-822-337 Light Neutral
19-822-338 Silver
19-822-339 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-822-337 Supplier Justrite™ Supplier No. 883027
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 19 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Self-Closing Doors is fully compliant and designed for safe storage of solvents and flammables in laboratories.
  • Under Fume Hood Cabinets are fully compliant and designed for the safe storage and venting of solvents and flammable liquids
  • Single-door cabinets are offered in either left or right-hinge door closures for easy operation where space is at a premium
  • Back wall vents with built-in flame arresters are positioned at lower left and top right for jurisdictions that mandate venting of vapors
  • Designed to meet OSHA and NFPA standards; self-close door styles also meet NFPA 1 and the International Fire Code
  • Constructed of sturdy 18-gauge (1-mm) thick double-wall, welded steel with 1-1/2 in. (38-mm) of insulating air space for fire resistance
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof 2 in. (51-mm) bottom sump. They adjust on 3 in. (76-mm) centers for versatile storage. Cabinets also feature dual vents with flame arresters, four adjustable self-leveling feet, grounding connector, and trilingual warning label. Durable lead-free epoxy/polyester powder-coat finish provides excellent chemical resistance
  • Self-close door(s) shuts and latches automatically when a fusible link melts at 165°F (74°C) under fire conditions. Unique, concealed self-closing mechanism offers obstruction-free access to contents
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Load Bearing Capacity 19 gal.
Color Light Neutral
Door Style Self-close