Learn More
HexArmor™ 19 in. Sleeve
Catalog No. 191305552 Shop All Performance Fabrics, Inc Products
Click to view available options
Size:
Small
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-5552 Large
19-160-613 Small
19-130-5553 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-130-5552 Supplier HexArmor™ Supplier No. AS019SL 9
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Size Large
Construction Light weight
Material SuperFabric™
Style Ambidextrous
Cut Resistance Rating Level 5
For Use With (Application) Glass, Metal and Wood handling, assembly, food packing and processing and animal handling
Length (Metric) 48.26 cm