Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

19-Maleimido-17-oxo-4,7,10,13-tetraoxa-16-azanonadecanoic Acid 95.0+%, TCI America™
SDP

Catalog No. M2666100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2666100MG 100 mg
M266625MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2666100MG Supplier TCI America Supplier No. M2666100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1263045-16-4
Molecular Formula C18H28N2O9
Molecular Weight (g/mol) 416.43
MDL Number MFCD13184954
InChI Key IAJVEYLYYHOZEY-UHFFFAOYSA-N
Synonym 1-Maleimido-3-oxo-7,10,13,16-tetraoxa-4-azanonadecan-19-oic Acid
PubChem CID 51340949
IUPAC Name 1-[3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido]-3,6,9,12-tetraoxapentadecan-15-oic acid
SMILES OC(=O)CCOCCOCCOCCOCCNC(=O)CCN1C(=O)C=CC1=O

Specifications

Quantity 100 mg
Formula Weight 416.43
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 19-Maleimido-17-oxo-4,7,10,13-tetraoxa-16-azanonadecanoic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.