Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BASIC™ Chemical Absorbent Pads

Catalog No. 19000672 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
17 gal.
20 gal.
34 gal.
Weight Capacity:
Heavyweight
Lightweight
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Weight Capacity
19-000-672 20 gal. Heavyweight
19-000-674 17 gal. Lightweight
19-000-673 34 gal. Lightweight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-000-672 Supplier Brady™ Supplier No. BPH100
Add to Cart
Add to Cart

Ideal spill response for chemicals like: citric acid, hydrofluoric acid, sodium hydroxide and many other chemicals.

  • Single Ply Meltblown Polypropylene construction
  • Chemically inert, making it ideal for use with aggressive fluids
  • Unique color helps to set these absorbents apart for proper disposal
  • Perforated to help reduce waste
  • Dimpled for added durability and reduced linting

Specifications

Color Yellow
Capacity (English) 20 gal.
Material Polypropylene
Type Chemical Absorbent Pad
For Use With (Application) Absorbent for applications with hazardous chemicals
Weight Capacity Heavyweight
Dimensions (L x W) 17 x 15 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.