Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ N100 Comfort Plus 8233 Particulate Respirator

Catalog No. 19002711 Shop All 3M Products
Change view
Click to view available options
N100; Faceseal; Welding; Not oil resistant
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-002-711 N100; Faceseal; Welding; Not oil resistant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Catalog No. 19-002-711 Supplier 3M™ Supplier No. 7000002028
Add to Cart
Add to Cart
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

N100 particulate respirator is designed to help provide reliable protection of at least 99.97 percent filtration efficiency against certain non-oil based particles.

  • NIOSH's highest rated filtration efficiency in a disposable respirator N100
  • Cool Flow™ exhalation valve reduces heat build-up inside the respirator
  • Compatible with a variety of protective eyewear and hearing protection
  • Adjustable noseclip helps provide a custom secure seal
  • Individually packaged
  • Advanced Electrostatic Media is designed for ease of breathing
TRUSTED_SUSTAINABILITY

Specifications

Aerosol Type N - Not Resistant to Oil
Assigned Protection Factor (APF) 10
Certifications (NIOSH) NIOSH 42 CFR 84, N100, approval number: TC-84A-1298
Certifications (OSHA) OSHA 1910.134
Color White
Design Type Cup/Molded
Disposable Single-use
Filter Type N100
Material Polypropylene, Polyester
Product Type General Purpose Particulate Respirator
Size Standard
With Exhalation Valve Yes
Filter Efficiency 99.97%
For Use With (Application) Transportation, Construction, Mining, Oil & Gas, General Manufacturing
Latex Free Yes
Material Headstrap Polyisoprene, Cotton/Polyester
Model 8233
Nose Piece Material Aluminum
Nosepiece Type Nose clip
Packaging Type Individually packaged
Quantity 1 Ea.
Sterility Non-sterile
With Faceseal Yes
Strap Attachment Type Adjustable Buckle Straps
FDA Cleared Surgical Mask No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.