Learn More
Brady™ Velocity™ Compact Nylon Lockout Padlocks, Keyed Different
Catalog No. 19029917 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-029-917 White
19-029-890 Black
19-029-894 Blue
19-029-898 Brown
19-029-902 Green
19-029-906 Orange
19-029-909 Purple
19-029-913 Red
19-029-921 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-029-917 Supplier Brady™ Supplier No. CPPLWHT25ALKD
Add to Cart
Add to Cart
Everything you love about Velocity™ Lockout Padlocks in a compact design.
Ideal for lockout applications where clearance or accessibility is limited.
Specifications
Color White
Key Type Keyed Different
Material Nylon
Quantity 1 Ea.
Product Type Safety Padlock
Height (English) Shackle 1 in.