Learn More
3M™ Tour-Guard™ V Protective Eyewear
Catalog No. 19039590 Shop All 3M Products
Change view
Click to view available options
No. per Pack:
100/Pk.
Dispenser Box (20/Box)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
19-039-590 Dispenser Box (20/Box)
19-039-591 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-039-590 Supplier 3M™ Supplier No. 7100089472
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ Tour-Guard™ V Protective Eyewear offers impact protection, integral sideshields and flexible, polycarbonate construction with over-the-glass coverage.
  • Economical protective eyewear serves needs of site visitors or general use
  • Wraparound side shields help provide extra protection
  • Medium size fits over most personal prescription eyewear
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380 nm
  • 3M Catalog# TGV01-100 , 3M Catalog# TGV01-20
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating UV-treated
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Clear
Product Type Safety Glasses
Frame Color Clear
Model TGV01-20
No. per Pack Dispenser Box (20/Box)
Temple Style Classic
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Frameless