Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell Kimtech™ Pure M5 Pleat-Style Cleanroom Face Masks
BIOPHARMAGREENER_CHOICE

Catalog No. 19039690 Shop All Ansell Products
Change view
Click to view available options
Fastening Type:
Earloops
Ties
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fastening Type
19-039-690 Ties
19-062-279 Earloops
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-039-690 Supplier Ansell Supplier No. 62742
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Three-layer, pleat-style face masks for use in ISO Class 5 and higher cleanroom environments. Available with earloops or soft ties.

  • Three-layer design
  • Made with polyester and cellulose
  • Pleat-style masks available with earloops or soft ties
  • Fully enclosed bendable nosepiece
  • Popular for biotech, medical device, electronics and telecommunication environments
  • Ultrasonically bonded ties are durable
  • Double bagged and recommended for ISO Class 5 and higher cleanroom environments
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Face Mask
Color Blue
Fastening Type Ties
Material Polyester
Size Standard
Sterility Non-sterile
Style Pleated
Cleanroom Class Class 100 (ISO 5)
Construction 3-ply
Disposable Single-use
Filter Efficiency 99%
For Use With (Application) Biotech, Medical Devices, Electronics, Telecommunications
Model 62742
Packaging Type Double-bagged
Series M5
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.