Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Vis-Tech™ Cut Resistant Glove with Nitrile Coating

Catalog No. 19048793 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-048-793 Large
19-048-790 X-Small
19-048-791 Small
19-048-792 Medium
19-048-794 X-Large
19-048-795 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-048-793 Supplier Wells Lamont™ Supplier No. Y9296L
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Vis-Tech™ Cut Resistant Glove with Nitrile Coating offers ANSI A4 cut resistance and an excellent grip, especially in wet and oily conditions. Features a high visibility color for safety and is lightweight and breathable.

  • Sandy nitrile coating on palm provides improved grip and protection in dry, wet, and oily conditions
  • Manufactured with a unique blend of high performance fibers and stainless steel to achieve a high level of comfort and cut resistance
  • Open, uncoated back circulates air, keeping hands to cool and comfortable
  • High visibility orange shell promotes safety
  • Launderable for a longer wear life
  • ANSI Cut Level 4, A4

Specifications

Coating Material Nitrile Foam
Color Yellow
Cut Resistance Rating ANSI A2
Hand Specific
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) General Assembly, Automotive Industry, Small Parts, and Glass Handling
Glove Cuff Style Knit
Series Y9256
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.