Learn More
3M™ Deluxe Headgear
Catalog No. 19053165 Shop All 3M Products
Change view
Click to view available options
Headgear, Deluxe
Catalog No.
19-053165 Headgear, Deluxe
1 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-053165 Supplier 3M™ Supplier No. 7000002290
Add to Cart
Add to Cart
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Face protection without snaps, bands or cams
Compact window provides more chest and shoulder clearance; large window provides maximum coverage.
  • Protects workers in metalworking, utilities, forestry and many other industries
  • High-strength thermoplastic crown protectors provide impact resistance over a wide range of temperatures
  • Flexible system lets users choose headgear and windows separately — allows window replacement in seconds
  • Precision ratchet adjustment for comfortable, snug fit; five-position crown strap for extra support
  • Floating window suspension distributes and reduces shock of impact
  • 3M Catalog# 82501-00000
Order Info
Replacement windows are available separately (Cat. Nos. 19-040-038, 19-060-701, 19-071-158, 19-053-169).
Compliance
Meets ANSI Z87.1-2003 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2003
For Use With (Equipment) Metalworking, utilities, forestry, and many other industries
Model 82501-00000