Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 50 Hooded Coverall

Catalog No. 19065501E Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Size (English) Chest:
33.25 to 37.25 in.
37.25 to 40.75 in.
39.75 tp 33.25 in.
40.75 to 44.25 in.
44.25 to 47.75 in.
47.75 to 51.25 in.
51.25 to 54.75 in.
54.75 to 58.25 in.
57.75 to 61.25 in.
Height (English):
5 to 5.58 ft.
5.22 to 6.16 ft.
5.25 to 5.58 ft.
5.41 to 5.75 ft.
6 to 6.33 ft.
6.16 to 6.33 ft.
6.33 to 6.58 ft.
6.58 to 6.83 ft.
6.75 to 7.08 ft.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (English) Chest Height (English)
19-065-501E 2X-Large 44.25 to 47.75 in. 6 to 6.33 ft.
19-065-501A Small 39.75 tp 33.25 in. 5 to 5.58 ft.
19-065-501B Medium 33.25 to 37.25 in. 5.25 to 5.58 ft.
19-065-501C Large 37.25 to 40.75 in. 5.41 to 5.75 ft.
19-065-501D X-Large 40.75 to 44.25 in. 5.22 to 6.16 ft.
19-065-501F 3X-Large 47.75 to 51.25 in. 6.16 to 6.33 ft.
19-065-501G 4X-Large 51.25 to 54.75 in. 6.33 to 6.58 ft.
19-065-501H 5X-Large 54.75 to 58.25 in. 6.58 to 6.83 ft.
19-065-501I 6X-Large 57.75 to 61.25 in. 6.75 to 7.08 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Should not be used around heat, flame, sparks or potentially flammable or explosive environments.
Catalog No. 19-065-501E Supplier DuPont™ Supplier No. NB122SWH2X002500
Add to Cart
Add to Cart
This item is not returnable. View return policy
Should not be used around heat, flame, sparks or potentially flammable or explosive environments.

Microporous film coveralls provide protection from non-hazardous aerosols, light liquid splash, and dry particles.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material ProShield
Color White
Disposable Single-use
Ankle Style Attached Boots, Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Standard, Elastic Face
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Model NB122S
Size (English) Chest 44.25 to 47.75 in.
Size (English) Inseam 28.25 in.
Size (Metric) Inseam 71.75 cm
Thickness 10 mil
For Use With (Application) Janitorial, Sanitation, General Industrial, Maintenance
Length (English) Sleeve 35.75 in.
Length (Metric) Sleeve 90.18 cm
Width (English) Chest 28.75 in.
Width (Metric) Chest 73.02 cm
Height (English) 6 to 6.33 ft.
Height (Metric) 1.83 to 1.93 m
Quantity 25/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.