Learn More
Bullard™ Loose Fitting Facepieces for PA20, PA30, PA30IS, EVA, EVAHL PAPRs
SDP
Catalog No. 19098042 Shop All Bullard Company Products
Click to view available options
Includes:
Contoured-fit, blue twist lock connection
Narrow profile, black quick-attach twist lock connection
Narrow profile, blue twist lock connection
Narrow profile, green twist lock connection
Traditional fit, Quick-attach twist lock connection
Size:
Small
Medium
Large
X-Large
Loose fitting facepieces provide comfortable and effective protection from a wide range of respiratory hazards.
  • Loose fitting facepiece
  • Head covers made of Tychem
  • Sewn-in headband suspension
  • Twist-lock breathing tube connection
  • For use with PAPR and SAR systems
  • Choose from a variety of sizes and color connections
  • Acetate lens material
  • NIOSH approved
Specifications
Assigned Protection Factor (APF) 25
Product Type Loose Fitting Facepiece
For Use With (Application) Pharmaceutical manufacturing, general industry and maintenance, woodworking, spray foam, painting, healthcare
For Use With (Equipment) Pa20, pa30, pa30is, eva™, eva™hl powered air purifying respirator
Includes Contoured-fit, blue twist lock connection
Latex Free Yes
Model 20LF2L50
Packaging Type Bulk Pack
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.