Learn More
Fisherbrand™ Siteliner Safety Glasses, Blue frame
Unilens design offers wide field of vision and exceptional clarity
 View more versions of this product
Catalog No. 19-130-2096
This item is not returnable. View return policy
Description
Description
Unilens design offers wide field of vision and exceptional clarity
  • Lightweight, impact-resistant nylon frames
  • One-piece, clear hardcoat polycarbonate lens for maximum protection
  • Scratch resistant; absorb >99.9% of harmful UV light
  • Molded-in sideshields and browguards for added protection against UV, dust and flying particles
  • Adjustable temples and lens inclination
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Specifications
Hardcoat
Clear
Polycarbonate
Meets ANSI Z87.1-2003 standards.
Black
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.