Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ Optime™ 95 Ear Muffs

Catalog No. 19145412 Shop All 3M Products
Change view
Click to view available options
Model:
H6A/V
H6B/V
H6F/V
XH6B/V
Style:
Behind-the-head
Over-the-head
Noise Reduction Rating (NRR):
19 dB
21 dB
Foldable:
No
Yes
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Style Noise Reduction Rating (NRR) Foldable
19-145-412 H6A/V Over-the-head 21 dB No
17-388B H6F/V Over-the-head 21 dB Yes
17-388A H6B/V Behind-the-head 21 dB No
19-808-022 XH6B/V Behind-the-head 19 dB Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-145-412 Supplier 3M™ Supplier No. 7000009667
Add to Cart
Edge
Add to Cart

PELTOR™ Optime™ 95 Ear Muffs are lightweight and comfortable and protect from noise levels up to 95 dBA.

  • Suitable for use when assessed noise levels reach up to 95dBA
  • Slim design for high comfort and continuous wear
  • Behind-the-Head style designed to be worn with most welding helmets
  • Folding style allows convenient storage
  • Over-the-Head style fits well over most safety equipment
  • Beige

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Beige
ANSI S3.1974 Yes
CSA Z94.2 Class B
Cushion Color Black
For Use With (Equipment) Fit well with most helmets, eyewear and other safety equipment
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
High Visibility No
Model H6A/V
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 21 dB
Product Type PELTOR™ Optime™
Headband Color Black
Dielectric No
Style Over-the-head
Foldable No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.