Learn More
3M™ Skull Screws™ Ear Plugs
Catalog No. 191501567 Shop All 3M Products
Change view
Click to view available options
Model:
P1300
P1301
Corded:
Corded
Uncorded
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Corded
19-150-1567 P1301 Corded
19-150-1566 P1300 Uncorded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-150-1567 Supplier 3M™ Supplier No. 7000127181
Add to Cart
Edge
Add to Cart
Skull Screws™ ear plugs metallic style and color makes them look solid, but they are made from ultra-soft foam to provide all-day comfort.
  • Tough look of threaded metal
  • Comfortable foam tips for extended wear
  • Cutting-edge stylish look may increase worker compliance in wearing hearing protection
  • Push-to-fit insertion style for easy fitting
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
Cord Color Gray
Cord Material Vinyl
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High Visibility No
Model P1301
Packaging Type Poly bag
Pairs per Package 480 Pairs/Cs
Plug Material Foam, thermoplastic
Plug Color Gray
Noise Reduction Rating (NRR) 32 dB
Corded Corded
Plug Shape Cone
Disposable Reusable
Metal Detectable No