Learn More
Pyramex™ Intruder™ Safety Eyewear
Catalog No. 19155581 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Amber
Blue
Blue, Mirror
Clear
Gray
Indoor/Outdoor, Mirror
Silver, Mirror
Frame Color:
Amber
Black
Blue
Clear
Clear with full foam padded frame
Clear with uncoated lens
Gray
Red
Silver
Lens Coating:
Anti-fog, UV-treated
UV-treated
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Frame Color Lens Coating
19-155-581 Blue, Mirror Blue UV-treated
19-155-571 Clear Clear Anti-fog, UV-treated
19-155-572 Clear Clear with full foam padded frame UV-treated
19-155-573 Clear Clear with uncoated lens -
19-155-574 Gray Gray UV-treated
19-155-576 Gray Gray Anti-fog, UV-treated
19-155-577 Gray Clear with full foam padded frame UV-treated
19-155-578 Amber Amber UV-treated
19-155-579 Blue Blue UV-treated
19-155-582 Indoor/Outdoor, Mirror Clear UV-treated
19-155-584 Indoor/Outdoor, Mirror Clear Anti-fog, UV-treated
19-155-585 Indoor/Outdoor, Mirror Clear with full foam padded frame UV-treated
19-155-715 Clear Black -
19-155-968 Clear Blue -
19-155-977 Clear Red -
19-155-580 Silver, Mirror Silver UV-treated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-155-581 Supplier Pyramex™ Supplier No. S4175S
Add to Cart
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Integrated nosepiece, lightweight, frameless protection
  • Lightweight, frameless protection
  • Superior comfort and fit
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Lens Coating UV-treated
    Certifications/Compliance ANSI Z87.1
    Lens Tint Blue, Mirror
    Frame Color Blue
    Lens Material Polycarbonate
    Frame Material Polycarbonate