Learn More
Texwipe™ AlphaMop™ Isolator Cleaning Tool
SDP
Catalog No. 191600022 Shop All Texwipe Products
Change view
Click to view available options
Product Type:
Cleanroom Mop
Mop
Replacement Mop Covers
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-160-0022 Mop
19-139-693 Replacement Mop Covers
19-139-692 Cleanroom Mop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-160-0022 Supplier Texwipe™ Supplier No. TX7101
Add to Cart
Add to Cart
Includes
Two handles (18 and 24 in. long), one polyester foam pad, and six TechniCloth™ polyester mop covers. Mop with telescoping handle includes six polyester mop covers.
Excellent for cleaning isolators, laminar flow hoods, biosafety cabinets, and glove boxes
  • Low-profile mop with rounded corners and swivel head to reach difficult-to-access areas
  • Made from autoclavable Celcon™ plastic
  • Replaceable foam pad ensures that mop cover conforms to surfaces being cleaned
Specifications
Product Type Mop
Autoclavable Autoclavable
For Use With (Application) For Cleanroom Class 4 to 8
Material Plastic