Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ Green 11 mil Nitrile Chemical Resistant Gloves, Unlined

Catalog No. 19162727 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-162-727 Medium
19-162-726 Small
19-162-729 X-Large
19-162-730 2X-Large
19-162-728 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-162-727 Supplier PIP™ Supplier No. 50N110G/M
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Unsupported, unlined nitrile gloves with raised diamond grip are ideal for light chemical applications, printing, meat and poultry processing.

  • Lightweight construction and hand specific design provide real comfort with protection
  • Provides flexibility, tactile sensitivity and dexterity sometimes lacking in heavier gauge unsupported nitriles
  • Protection from chemical and physical hazards not always found in thinner gauge nitrile disposables
  • Unlined to reduce potential of product contamination
  • Straight cuff adds additional length to protect the forearm from liquid run-off
  • Raised Diamond pattern for an enhanced grip
  • Chlorinated to harden and cleanse the surface of the glove
  • Made from component materials which comply with Federal regulations for food contact, 21 CFR, 170-199

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) CE, EN 374-3
Chlorinated Yes
Applications Industrial
Color Green
Finger Thickness (English) 11 mil
Finger Thickness (Metric) 0.28 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 11 mil
Palm Thickness (Metric) 0.28 mm
Product Type Chemical Protection Gloves
Size Medium
Thickness (English) 11 mil
Thickness (Metric) 0.27 mm
EN388 Standard EN 388:3001
EN 421 No
EN ISO 374-1 CHEMICAL LETTER CODES ABK
For Use With (Application) Light chemical applications, printing, meat and poultry processing
Glove Cuff Style Straight
Grip Design Raised Diamond
Lining Unlined
Model 50-N110G/M
Series Assurance™ 50-N110G
Length (English) 13 in.
Length (Metric) 33 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.