Learn More
Vileda Professional™ DuoPress™ Wringer
Catalog No. 19200830 Shop All Vileda Professional Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-200-830 1 Ea.
1 options
Catalog No. 19-200-830 Supplier Vileda Professional™ Supplier No. 168207
Add to Cart
Add to Cart
The CE Trolley range has been specifically designed for use in controlled environments where maximum contamination control is needed. This DuoPress™ wringer is part of the CE Bucket Trolley line for wringing of flat mops.
  • For use on the CE Bucket Trolley with the bucket method
  • 100% polypropylene chassis and stainless steel lever
  • Effectively wrings CE Duo™ mops
  • Ergonomic lever for maximum pressure with minimal effort
  • Base of press support dropped 10 mm to create additional space for mops, making it easier to wring
Specifications
Product Type Mop Wringer
Handle Material Polypropylene and Stainless steel
Includes Wringer base, Support plate and Wringer
Quantity 1 Ea.