Learn More
Moldex™ Probed Cartridge Kit for 7000/9000 Series Respirators
Kit includes everything needed to perform quantitative fit testing using TSI Portacount on all 8000 Series Moldex™ respirators.
Supplier: Moldex™ 7006
Description
- Comply with OSHA standards for quantitative fit testing
- Includes:
- 1 Probed Cartridge For Use With 7000 or 7800 Series
- 1 Probed Cartridge For Use With 9000 Series
- 1 Pair 7940 P100 Filter Disks
- 1 Pair 7920 Piggyback Adapters
- Instructions and certificates
- Respirators sold separately
Compliance
NIOSH approved.
Specifications
Probed Cartridge Kit | |
No | |
7006 |