Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ EcoPolyBlend™ Drum Management System
GREENER_CHOICE

Catalog No. 19413816 Shop All Justrite Safety Group Products
Change view
Click to view available options
Type:
Dispensing Shelf
Drum Management Base Module
Drum Management Stack Module
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
19-413-816 Drum Management Stack Module
19-413-815 Drum Management Base Module
19-413-820 Dispensing Shelf
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-413-816 Supplier Justrite™ Supplier No. 28669
Add to Cart
Add to Cart

Features a space-saving stackable design

100% molded polyethylene construction provides maximum chemical resistance, preventing corrosion, deterioration and leaks

  • Stores up to four 110 and/or 200L (30 and/or 55 gal.) drums
  • Stores drums of Class 1B or 1C flammables and combustibles or nonhazardous liquids
  • Consists of three parts: a base module, stack module and dispensing shelf
  • Molded-in drum lock-channels hold drums in place

Base Module

  • Holds two drums with a 1388kg (3060 lb.) total load capacity
  • Easily relocates with an empty forklift
  • 250L (66 gal.) open-view containment sump makes it easy to spot any spills for a quick cleanup
Stack Module
  • Holds two additional drums to double the system capacity for a load of 680kg (1500 lb.)
  • When storing both 110 and 200L (30 and 55-gal.) drums, the smaller drums should be placed on the top of the stack module
Dispensing Shelf
  • Mounts securely to the stack module for convenient dispensing from the upper level
  • Has a load capacity of 27 kg (60 lb.)

Compliance

EPA, NFPA, OSHA, SPCC

Specifications

Certifications/Compliance SPCC, EPA, NFPA, OSHA
Color Black
Material Polyethylene
Type Drum Management Stack Module
For Use With (Application) Efficient storage, positive spill control, liquid dispensing
Holds 4 Drums
Load Bearing Capacity (English) 1500 lb.
Load Bearing Capacity (Metric) 680.4 kg
Width (English) 49 in.
Width (Metric) 124.46 cm
Height (English) 12 in.
Height (Metric) 30.4 cm
Depth (English) 46 in.
Depth (Metric) 116.8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.