Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tert-Butyl(Chloro)Diphenylsilane

Catalog No. 195537250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-553-7250G 250 g
19-553-71KG 1 kg
19-553-710G 10 g
19-553-750G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 19-553-7250G Supplier Sigma Aldrich Supplier No. 195537250G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silylating reagent used in the protection of alcohols and the preparation of silyl ethers.

Specifications

CAS 58479-61-1
Density 1.057 g/mL at 25°C
Boiling Point 90°C (0.01 mmHg, lit.)
Molecular Formula C16H19ClSi
Refractive Index n20/D 1.568
Linear Formula (CH3)3CSi(C6H5)2Cl
MDL Number MFCD00000497
Quantity 250 g
Synonym tert-Butyldiphenylchlorosilane; TBDPSCl
Molecular Weight (g/mol) 274.86 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.