Learn More
ALADDIN SCIENTIFIC CORPORATION Melamine, 100g
Supplier:  ALADDIN SCIENTIFIC CORPORATION M108433100G
Encompass_Preferred
Melamine can be used as a- Reactant in the preparation of melamine trisulfonic acid (MTSA) which is used as a catalyst in various reactions Nitrogen source for the large scale synthesis of nitrogen-doped graphene (NG) Heterogeneous catalyst in combination with lewis acid ZnI2 for conversion of carbon dioxide to cyclic carbonates
Catalog No. 50-000-82515
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.