Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetriol 98.0+%, TCI America™
SDP

Catalog No. C227525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C227525G 25 g
C22755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C227525G Supplier TCI America Supplier No. C227525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50409-12-6
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00043062
InChI Key FSDSKERRNURGGO-UHFFFAOYSA-N
PubChem CID 230351
IUPAC Name cyclohexane-1,3,5-triol
SMILES C1C(CC(CC1O)O)O

Specifications

Melting Point 188°C
Color White
Quantity 25 g
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetriol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.