Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1E)-1-tert-Butoxy-1-(trimethylsilyloxy)propene 92.0+%, TCI America™
SDP

Catalog No. B32981G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32981G 1 g
B32985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32981G Supplier TCI America Supplier No. B32981G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 72658-10-7
Molecular Formula C10H22O2Si
Molecular Weight (g/mol) 202.369
InChI Key OJYHIAHTMZIVKY-UHFFFAOYSA-N
PubChem CID 57353179
IUPAC Name trimethyl-[1-[(2-methylpropan-2-yl)oxy]prop-1-enoxy]silane
SMILES CC=C(OC(C)(C)C)O[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 53°C
Quantity 1 g
UN Number 1993
Formula Weight 202.37
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material (1E)-1-tert-Butoxy-1-(trimethylsilyloxy)propene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.