Learn More
MSC Made in USA SOF-064 1" ID x 1-1/4" OD, 1/8" Wall Thickness, Cut to Length (50' Standard Length) PVC Tube Clear, 28 Max psi, 68 Shore A Hardness
Supplier:  MSC 79814687
Encompass
This item must be ordered in multiples of 25 per supplier requirements
Made in USA SOF-064 1" ID x 1-1/4" OD, 1/8" Wall Thickness, Cut to Length (50' Standard Length) PVC Tube Clear, 28 Max psi, 68 Shore A Hardness
Catalog No. NC1836129
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.