Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,2H,2H-Nonafluorohexyl Iodide 99.0+%, TCI America™
SDP

Catalog No. P115525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P115525G 25 g
P11555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P115525G Supplier TCI America Supplier No. P115525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2043-55-2
Molecular Formula C6H4F9I
Molecular Weight (g/mol) 373.988
MDL Number MFCD00039409
InChI Key CXHFIVFPHDGZIS-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorohexyl iodide, 2-perfluorobutyl ethyl iodide, hexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo, 1h,1h,2h,2h-nonafluorohexyl iodide, 1-iodo-1h,1h,2h,2h-nonafluorohexane, 1,1,2,2-tetrahydroperfluorohexyliodide, 2-nonafluorobutyl ethyl iodide, 1-iodo-1h,1h,2h,2h-perfluorohexane, 1h,1h,2h,2h-perfluoro-1-iodohexane, 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodo-hexane
PubChem CID 74887
IUPAC Name 1,1,1,2,2,3,3,4,4-nonafluoro-6-iodohexane
SMILES C(CI)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point -25°C
Color Red
Boiling Point 126°C
Quantity 25 g
Formula Weight 373.99
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,2H,2H-Nonafluorohexyl Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.