Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,5H-Octafluoropentyl 1,1,2,2-Tetrafluoroethyl Ether 98.0+%, TCI America™
SDP

Catalog No. O042225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O042225G 25 g
O04225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O042225G Supplier TCI America Supplier No. O042225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16627-71-7
Molecular Formula C7H4F12O
Molecular Weight (g/mol) 332.089
MDL Number MFCD00155904
InChI Key ZNBGTBKGFZMWKR-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5-Octafluoropentyl 1,1,2,2-Tetrafluoroethyl Ether
PubChem CID 2782579
IUPAC Name 1,1,2,2,3,3,4,4-octafluoro-5-(1,1,2,2-tetrafluoroethoxy)pentane
SMILES C(C(C(C(C(F)F)(F)F)(F)F)(F)F)OC(C(F)F)(F)F

Specifications

Color Colorless
Boiling Point 133°C
Quantity 25 g
UN Number 3271
Formula Weight 332.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,5H-Octafluoropentyl 1,1,2,2-Tetrafluoroethyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.