Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,7H-Dodecafluoro-1-heptanol, 97%

Catalog No. AAB2014422
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2014422 100 g
AAB2014414 25 g
AAB2014436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2014422 Supplier Thermo Scientific Chemicals Supplier No. B2014422
Add to Cart

CAS: 335-99-9 | C7H4F12O | 332.089 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-99-9
Molecular Formula C7H4F12O
Molecular Weight (g/mol) 332.089
MDL Number MFCD00039630
InChI Key BYKNGMLDSIEFFG-UHFFFAOYSA-N
Synonym 1h,1h,7h-dodecafluoro-1-heptanol, 1,1,7-trihydroperfluoroheptanol, 1h,1h,7h-dodecafluoroheptanol, 1-heptanol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro, unii-lpx5k7ec3w, 1,1,7-trihydrododecafluoroheptanol, 1,1,7-trihydroperfluoroheptyl alcohol, 1-heptanol, 1h,1h,7h-dodecafluoro, 1,1,7-trihydrododecafluoroheptan-1-ol, 1h,1h,7h-dodecafluoro-1-hydroxyheptane
PubChem CID 67639
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptan-1-ol
SMILES C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point -20°C
Density 1.762
Boiling Point 169°C to 170°C
Flash Point None
Refractive Index 1.318
Quantity 100 g
Beilstein 1800110
Formula Weight 332.09
Percent Purity 97%
Chemical Name or Material 1H,1H,7H-Dodecafluoro-1-heptanol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-406-6
RTECSNumber MJ4500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.