Learn More
Sigma Organic Chemistry 1H-Pyrazole-3-carbaldeh | 50mg | MFCD00129925
Supplier: Sigma Organic Chemistry CDS00991850MG

1H-Pyrazole-3-carbaldeh , 50mg
About this Item:
MDL #: MFCD00129925
Molecular Weight: 96.09
UNSPSC Code: 12352200
Chemical Formula: C4H4N2O
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.