Learn More
Sigma Organic Chemistry 1H-Pyrazole-4 5-diamine | 1G | MFCD09473558
Supplier: Sigma Organic Chemistry CBR021171G

1H-Pyrazole-4 5-diamine , 1G
About this Item:
MDL #: MFCD09473558
Molecular Weight: 187.24
UNSPSC Code: 12352200
Chemical Formula: C11H13N3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.