Learn More
Sigma Organic Chemistry 1H-Pyrazole-5-boronic p | 50mg | MFCD09743409
Supplier: Sigma Organic Chemistry CDS01259950MG

1H-Pyrazole-5-boronic p , 50mg
About this Item:
MDL #: MFCD09743409
Molecular Weight: 194.04
UNSPSC Code: 12352200
Chemical Formula: C9H15BN2O2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.