Learn More
Sigma Organic Chemistry 1H-Pyrrolo 2 3-bpyriDNe | 100mg | MFCD06659752
Supplier: Sigma Organic Chemistry CDS009698100MG
1H-Pyrrolo 2 3-bpyriDNe , 100mg
About this Item:
MDL #: MFCD06659752
Molecular Weight: 162.15
UNSPSC Code: 12352200
Chemical Formula: C8H6N2O2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.